EMS Hobby Blade and Handle, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71982-HD 71982
103301-802BX 6.9 CAD
103301-802 103302-328
EMS Hobby Blade and Handle, Electron Microscopy Sciences
Razors and Blades
Hobby blades are used in a wide range of applications due to the simple but precise cutting tip.
These 0.021" carbon steel blades fit most standard handles
Handles are made from aluminum construction for durability and they have a 4 jaw check which holds the blade securely.
They are precision ground for sharpness and durability.