Pierce™ UltraLink™ Immobilized Protein G, Thermo Scientific
Supplier: Thermo Scientific
53125 53126 53127 53128
CAPI53125EA 969.3 CAD
CAPI53125 CAPI53126 CAPI53127 CAPI53128
Pierce™ UltraLink™ Immobilized Protein G, Thermo Scientific
Protein Purification Protein Purification Resins
Protein G UltraLink™ Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein G has been immobilized for use in IgG purification methods
  • Immobilized Protein G – specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat
  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
It is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods.
Delivery information: Available as resin or as Protein G UltraLink Columns (2 x 2 mL resin pre-packed in columns with twist-off bottom tabs and an accessory pack containing two white tips).