Pierce™ Pegylation Reagents, Methyl-PEG-NHS-Maleimide, Thermo Scientific
Supplier: Thermo Scientific
22711
22713
CAPI22711EA
1162.83
CAD
CAPI22711
CAPI22713
Pierce™ Pegylation Reagents, Methyl-PEG-NHS-Maleimide, Thermo Scientific
Protein Purification
Protein Purification Reagents
Pierce™ MM(PEG)n reagents are methyl-terminated PEG compounds activated with a maleimide group for covalent pegylation of sulfhydryls on proteins or assay surfaces.
- Choice of 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application
- PEG spacer provides advantages that include increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Maleimide activated for efficient PEGylation of primary amines at pH 6.5 to 7.5; reaction of maleimide group results in formation of stable, irreversible amide bonds
- Easy to follow instructions increase the likelihood of a successful outcome