Tweezers WA1 Handle with 17-STD Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TIP-17-STD 10004-S17-SM
103302-292EA 18.57 CAD
103302-292 103302-600
Tweezers WA1 Handle with 17-STD Tips, Electron Microscopy Sciences
Forceps
These tweezers have an anti-magnetic WA1 handle with replaceable 17-STD tips.
  • Hand made of the highest quality steels and alloys
  • Tips attach with screws
  • High precision and perfectly finished to suit the application
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as inox.