Pepsin Reagent, Ready To Use, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
64142-05 64142-06
103303-060EA 190.11 CAD
103303-060 103305-696
Pepsin Reagent, Ready To Use, Electron Microscopy Science
Protein Extraction Protein Extraction Reagents
Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
  • Antigen retriever for some antibodies in IHC
  • Please refer to primary antibody protocol for IHC
Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC.
Pepsin digestion of FFPE tissue section improves accessibility of antibodies to tissue antigens.