Saws, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70566-32 70566-30 70566-31
103302-192EA 170.83 CAD
103302-192 103305-476 103306-032
Saws, Electron Microscopy Sciences
Saws
Three styles of manual bone saws are available.
  • Broad bladed Satterlee Saw
  • Thinner rounded top Charriere Saw
  • Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.