Section Block, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62710 62711 62712
103305-762EA 320.65 CAD
103305-762 103306-198 103306-942
Section Block, Electron Microscopy Sciences
Buffers Electrophoresis Buffers Blocking Buffers
Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.
Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.
Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.