Stainless Steel Drip Troughs, Interdyne Inc
Supplier: Interdyne
HDT-18-2 HDT-20-2 HDT-24-2 HDT-30-2 HDT-32-2 HDT-36-2 HDT-42-2 HDT-48-2 HDT-60-2
VF-HDT-18-2EA 229.1 CAD
VF-HDT-18-2 VF-HDT-20-2 VF-HDT-24-2 VF-HDT-30-2 VF-HDT-32-2 VF-HDT-36-2 VF-HDT-42-2 VF-HDT-48-2 VF-HDT-60-2
Stainless Steel Drip Troughs, Interdyne Inc
Trays/Dishes/Pots Spillage Trays/Pans
All stainless steel construction, made from 20 gauge material, Type 304 stainless steel.
  • 1" mounting flange
  • 3 ft flexible drain hose included
4 finish.