Mouse Recombinant RELM G (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-89-5UG 200-89-25UG 200-89-100UG 200-89-500UG 200-89-1MG
76051-342EA 133.4 CAD
76051-342 76051-344 76051-346 76051-348 76051-350
Mouse Recombinant RELM G (from E. coli)
Proteins and Peptides
Resistin-like molecule-gamma (RELM-γ) is a member of the RELM family of secreted proteins containing C-terminal cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-γ is secreted by peripheral blood granulocytes, bone marrow, spleen, intestine, and lung. RELM-γ functions to promote and regulate promyelocytic differentiation, in addition to regulating nutrient-associated insulin sensitivity in the intestinal tract. Rodents secrete all four RELM family members, whereas Resistin and RELM-β are the only RELM family members found in humans.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.