Mouse Recombinant IL-1 alpha (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-87AF-5UG 200-87AF-20UG 200-87AF-1MG 200-87AF-100UG 200-87AF-500UG
76051-292EA 142.6 CAD
76051-292 76051-294 76051-296 76051-298 76051-300 77660-878 77660-880
Mouse Recombinant IL-1 alpha (Animal free) (from E. coli)
Proteins and Peptides
Interleukin 1 alpha (IL-1α) is expressed by epithelial cells, activated macrophages, neutrophils, and endothelial cells to regulate immune responses. IL-1α signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter. IL-1α and the independently regulated IL-1β protein have overlapping proinflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.