Human Recombinant FGF-6 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-201-5UG 100-201-25UG 100-201-100UG 100-201-500UG 100-201-1MG
76050-892EA 123.67 CAD
76050-892 76050-894 76050-896 76050-898 76050-900
Human Recombinant FGF-6 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.