Human Recombinant FGF-5 (Animal Free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-200-100UG
100-200-500UG
100-200-1MG
100-200-10UG
100-200-50UG
100-200AF-100UG
76050-876EA
623.29
CAD
76050-876
76050-878
76050-880
76051-234
76051-236
77660-732
Human Recombinant FGF-5 (Animal Free) (from E. coli)
Proteins and Peptides
Fibroblast growth factor 5 (FGF-5) is a secreted heparin-binding growth factor that binds to FGF receptors 1 and 2 (FGFR1 and FGFR2). FGF-5 is expressed in the mesenchyme, skeletal muscles, central nervous system, and hair follicles to promote cell differentiation and proliferation. FGF-5 functions as a regulatory factor during hair elongation and skeletal muscle development.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.