Human Recombinant WISP-2 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-197-5UG 100-197-20UG 100-197-100UG 100-197-500UG 100-197-1MG
76050-912EA 133.4 CAD
76050-912 76050-914 76050-916 76050-918 76050-920
Human Recombinant WISP-2 (from E. coli)
Proteins and Peptides
WNT1-inducible-signaling pathway protein 2 (WISP-2) is a member of the CYR61/CTGF/NOV (CCN) family of regulatory factors. WISP-2 is expressed in ectodermal, mesodermal, and endodermal lineages, including primary osteoblasts, fibroblasts, mesenchymal stem cells, and adipogenic precursor cells. WISP-2 is a canonical WNT ligand that regulates cell proliferation, adhesion, and metastasis. Secreted WISP-2 promotes mesenchymal precursor cell proliferation and maintains them in an undifferentiated state. In bone-forming osteoblasts, WISP-2 promotes osteoblast adhesion and inhibits osteocalcin production.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.