Lacey Formvar with Carbon Film Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
LFC200-CU-50 LFC300-CU-50 LFC300-CU-100 LFC400-CU-50 LFC400-CU-100 LFC200-NI-50 LFC200-NI-100 LFC200-NI-150 LFC300-NI-50 LFC300-NI-100 LFC300-NI-150 LFC400-NI-50 LFC400-NI-150 LFC200-AU-100 LFC200-AU-150 LFC300-AU-50 LFC300-AU-100 LFC200-AU-50 LFC300-AU-150 LFC400-AU-50 LFC400-AU-100 LFC400-AU-150
103302-868EA 166.33 CAD
103302-868 103302-874 103302-876 103302-880 103302-882 103303-032 103303-034 103303-036 103303-038 103303-040 103303-042 103303-044 103303-048 103304-540 103304-542 103304-544 103304-546 103305-490 103305-492 103305-494 103305-496 103305-960
Lacey Formvar with Carbon Film Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Provides support without interfering with tranmission electron microscopy observation.
  • Copper, nickel and gold grids
  • Mesh grid sizes of 200, 300, and 400
  • Optically checked followed by batch testing in the EM
This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. Average opening sizes are 50, 100 and 150 microns. This type of pattern provides support without interfering with specimen observation.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.