Nunc™ Cell Culture EasYDishes, Thermo Scientific
Supplier: Thermo Fisher Scientific
150462 150464 150466 150468 150460
76169-584CS 368.3 CAD
76169-584 76169-586 76169-588 76169-590 76169-634
Nunc™ Cell Culture EasYDishes, Thermo Scientific
Culture Dishes Cell Culture Dishes
Nunc™ EasYDishes are the next generation of cell culture dishes that improve handling, stacking, and transporting of cell cultures in the lab.
  • Beveled grip ring makes it easier to handle and manage with a gloved hand
  • Proprietary notch design maximizes ventilation while maintaining sterility, and restricts lid movement to ensure secure lid-to-dish fitting
  • Raised outer edge on dish lid allows stable stacking during transportation of cell cultures
  • Orientation marks on dish bottom simplify cell location under the microscope
  • Writing area improves traceability
More Product Information