Sample Tubes, 4×300 ml, Büchi
Supplier: Buchi
037377 037377
89370-934EA 988.7 CAD
89370-934 CA89370-934
Sample Tubes, 4×300 ml, Büchi
Tubes Test Tubes
This sample tubes feature wider diameter, constricted condensation zone, thicker glass wall and can process higher volumes compared to sample tubes from other manufacturers.
  • Precision joint: tightness, avoid fume leaks
  • Constricted condensation zone; no splashing
  • Wider diameter; reduced foaming
  • Thicker glass wall, longer lifetime
The engineering and design ensures a better seal to the suction module of the digester and the splash protector of the distillation unit. All BUCHI Kjeldahl glassware (100, 300 and 500 ml) maintain the same integrity and upper opening shape. By use of built-in distillation unit design, all glassware will fit properly.