Float Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
456395 456398
76169-972PK 67.23 CAD
76169-972 76169-974
Float Racks for Microtubes, Globe Scientific
Racks Tube Racks
Autoclavable polypropylene construction for strength, buoyancy, and temperature resistance.
  • Racks float with full load of filled tubes
  • Molded handle in center of rack for comfort and support during transport
  • Legs allow rack to stand upright for use on benchtop
  • Use in ice, water baths, or on the benchtop
  • 65 mm round rack fits most 400 mL beakers
Commonly found in cloning, microbiology, and immunology labs.