Place Microscope Slide Files, 20, Globe Scientific
Supplier: Globe Scientific
513029W 513029G 513029B 513029A 513029Y
76169-996CS 457.27 CAD
76169-996 76169-998 76170-000 76170-002 76170-004
Place Microscope Slide Files, 20, Globe Scientific
Slide Holders
The durable 20-Place microscope slide file provides added protection to slides during routine storage.
  • Holds up to 20 microscope slides horizontally
  • Each space is pre-numbered for easy identification
  • Transparent cover protects slides and allows for easy reading of ID labels
  • Covers can be swung behind the base for ready access to the slides
  • Five color options for organization and classification of slides
The tray is produced from polystyrene (PS) and the lid is produced from styrene-acrylonitrile (SAN) which provides a longer usable life.