Cholesterol Fluorometric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
10007640-1 10007640-5
75816-698EA 392.82 CAD
75816-698 75816-700
Cholesterol Fluorometric Assay Kit, Cayman Chemical Company
Assays Protein Assays
For the sensitive quantitation of total cholesterol in plasma or serum.
  • Quantitation of total cholesterol in plasma or serum
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 530 – 540 nm, em 585 – 595 nm)
Cholesterol, particularly in the form of LDLs, is well understood to be associated with increased risk of coronary heart disease. The measurement of cholesterol is one of the most common tests performed in the clinical laboratory setting. However, simple and easy assays for cholesterol in the research lab have not been readily available. Cayman’s Cholesterol Assay provides a simple fluorometric method for the sensitive quantitation of total cholesterol in plasma or serum.