Prostaglandin D₂-MOX ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical Company
512011-480 512011-96
75817-268EA 2332.27 CAD
75817-268 75817-270
Prostaglandin D₂-MOX ELISA Kit, Cayman Chemical
Assays ELISAs
Competitive ELISA for quantitation of PGD2 in biological samples
  • Treatment of samples with MOX hydrochloride converts unstable PGD2 to stable PGD2-MOX
  • Highly sensitive
  • Measure PGD2-MOX levels down to 3.1 pg/ml
  • Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
  • Midpoint: 50% B/B0: 10-35 pg/ml
This PGD2-MOX ELISA is based on the conversion of PGD2 to a stable MOX derivative. Treatment of the sample with MOX HCl converts PGD2 into PGD2-MOX, preventing its further chemical degradation. The antiserum used in the assay was developed using conjugates of this derivative and is very specific for PGD2-MOX.