Syringe Filters, PTFE, Bonna-Agela Technologies
Supplier: Phenomenex
AS042520-P-19 AS043301
97011-456PK 1619.03 CAD
97011-456 97011-458 97011-460 10124-894
Syringe Filters, PTFE, Bonna-Agela Technologies
Filters Syringe Filters
These hydrophobic PTFE syringe filters have a broad chemical compatibility.
  • Low protein-binding
  • Low extractable
Order Now
SPECIFICATIONS
Environmentally Preferable