Motic BA310 Elite Standard Lab Phase Package
Supplier: Motic
1100100402645
10854-392EA 6142.31 CAD
10854-392
Motic BA310 Elite Standard Lab Phase Package
Microscopes
The BA310 Elite has been specifically designed to serve the daily routine needs of universities, clinics and labs.
  • 30W Koehler illumination yields a powerful and adjustable light source to fully expose even the weakest stained specimens
  • Hard coated/chemical resistant stage with improved slide holder grip and 76 x 50 travel range
  • High quality CCIS EC objectives provide fully corrected, perfected intermediate images without colored fringes
  • Multiple illumination options (Koehler 6V/30W halogen or 3W LED
  • Ergonomic eyepiece design for long periods of fatigue free viewing
  • High eyepoint eyepieces are perfect for glasses wearers and provide diopter adjustment for precision reticle use
  • Anti-fungus treatment is applied to this microscope to protect from fungus growth in high humidity environments
This phase package has outfited the BA310E with a variety of accessories to make it a highly capable phase microscopy instrument
This Phase Package includes all of the high quality components necessary to conduct routine professional clinic/lab phase microscopy work. Included in the package is Motic's BA310 Elite, which is an advanced upright compound microscope designed to be fully upgradable and withstand the demands of daily lab use. It has been outiftted with quality accessories for phase microscopy.
Users will enjoy the vivid and accurate images provided by the color corrected infinity optics, including the three phase objectives which come with the package. The high eyepoint eyepieces come with rubber eyecups for maximum comfort and minimal strain for the user. These eyepieces yield consistent diopter adjustment, enabling precise usage of reticles for measuring/counting.
The BA310 Elite's trinocular head provides the option of using a wide variety of cameras while being able to utilize both eyepieces. It also rotates and swivels a full 360° to provide maximum comfort for any user.
With special phase objectives, condenser and centering telescope, the BA310 Elite Phase Package is the ideal choice for routine professional phase microscopy.
Ordering information: Includes 45 mm blue filter, immersion oil (5 mL), power cord, allen hexagonal key, thumb screw, vinyl dust cover, and instruction manual. For other phase accessories, a phase centering telescope is available. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.