Half Size Wireless Nylon Tube Racks, Globe Scientific
Supplier: Globe Scientific
456845G 456845M 456845N 456845R 456845W 456845Y 456445B 456445G 456445M 456445N 456445R 456445W 456445Y 456545B 456545G 456545M 456545N 456545R 456545W 456545Y 456645B 456645G 456645M 456645N 456645R 456645W 456645Y 456745B 456745G 456745M 456745N 456745R 456745W 456745Y 456845B
76170-102EA 35.67 CAD
76170-102 76170-104 76170-106 76170-108 76170-110 76170-112 76170-204 76170-206 76170-208 76170-210 76170-212 76170-214 76170-216 76170-218 76170-220 76170-222 76170-224 76170-226 76170-228 76170-230 76170-232 76170-234 76170-236 76170-238 76170-240 76170-242 76170-244 76170-246 76170-248 76170-250 76170-252 76170-254 76170-256 76170-258 76170-260
Half Size Wireless Nylon Tube Racks, Globe Scientific
Racks Tube Racks
These fully autoclavable racks are molded from reinforced nylon for strength and durability.
  • Single piece, three tier grid design for secure and stable racking of tubes
  • Good temperature and chemical resistance to withstand a variety of lab procedures
  • Large, flat endplates for labeling and easy sample identification
  • Dense nylon material will not float in water bath
  • 15 ml conicals fit in 20 mm racks and 50 ml conicals fit in 30 mm racks
  • Seven colorful options to keep samples and benchtop organized
With no coatings to peel or chip, the wireless tube racks are a great alternative to wire coated racks where the formation of rust can be an issue.