Stacking/Connecting Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
456330C 456330B 456330Y 456330G 456330N 456335C 456335B 456335Y 456335G 456335N
76170-058EA 45.42 CAD
76170-058 76170-060 76170-062 76170-064 76170-066 76170-068 76170-070 76170-072 76170-074 76170-076
Stacking/Connecting Racks for Microtubes, Globe Scientific
Racks Tube Racks
These polycarbonate racks are provided with the ability to stack and connect your racks together with ease.
  • Autoclavable polycarbonate rack for sample processing and short to medium term storage down to –80 °C
  • Racks can be stacked or coupled with other racks to form a larger workstation
  • Numeric markings for sample identification
The racks are provided with a standard 2×12 orientation for easy view and recall of tube samples.