ZR BashingBead Lysis/Filtration Tubes, Zymo Research
Supplier: Zymo Research
S6010 S6011 S6012-50
76004-190EA 355.59 CAD
76004-190 76005-126 76005-128
ZR BashingBead Lysis/Filtration Tubes, Zymo Research
Tubes Test Tubes
The state of the art, ultra-high density beads are fracture resistant, chemically inert.
  • Capacity: 1000 µl
  • 0.1 and 0.5 mm in impact resistant 2 ml tube
Beads are ideal for tough-to-lyse bacteria, yeast, fungi and algae.