dNTP Mix, Zymo Research
Supplier: Zymo Research
D1000 D1005 D1010 D1015 D1020
76004-180EA 52.9 CAD
76004-180 76004-182 76005-226 76005-228 76005-230
dNTP Mix, Zymo Research
Nucleic Acid Reagents Nucleotides
The dNTP Mix, along with dATP, dTTP, dGTP, and dCTP mixes from Zymo Research are of ultra-high purity and can be used to generate DNA by PCR using ZymoTaq™ or other DNA polymerases.
  • Ready to use dNTP Mix (dATP, dTTP, dGTP, dCTP) of ultra high purity; >99% trisphosphate by HPLC
  • Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases
  • Free of endo-, exodeoxyribonuclease, ribonuclease, phosphatase, and nicking activities