SP Bel-Art Giant Polygon Spinbar Magnetic Stirring Bars with Pivot Ring, Bel-Art Products, a part of SP
Supplier: VWR International
74950-280 74950-282
74950-280EA 334.15 CAD
74950-280 74950-282
SP Bel-Art Giant Polygon Spinbar Magnetic Stirring Bars with Pivot Ring, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Designed for stirring large volumes in vessels such as drums and tanks
  • Marring of plastic containers is diminished
  • Multifaceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Ideal for use with Cool Stir® Large Volume Stirrer, H37028-0001; capable of stirring volumes up to 190 L (50 gallons)
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
  • Autoclave safe
A molded pivot ring minimizes the contact area between the bar and the vessel, thus reducing friction and chattering.