Backyard Brains® Plant Spikerbox
Supplier: BACKYARD BRAINS, INC.
BYBP0084
470231-880KT 296.8 CAD
470231-880
Backyard Brains® Plant Spikerbox
Data Loggers
Perform Plant Electrophysiology Experiments At Home Or In The Classroom!
  • Easy to use
  • Output to either a PC or a smartphone
Like previous inventions, the Plant SpikerBox is extremely easy to use and making it accessible for students as young as middle school. The device works by recording the electrical activity responsible for different plant behaviors. For example, the Venus Flytrap uses an electrical signal to determine if a prey in its trap; the SpikerBox reveals these invisible messages and allows you to visualize them on your mobile device. For the first time ever, you can peer into the fascinating world of plant signaling and plant behaviors.
Delivery information: Plant Spiker box includes conductive electrode gel, plant communciation cable, precise stimulation instruments, various length grounding / recording cables, smartphone and USB cable.