Autoclave Bags, Chemglass
Supplier: Chemglass
CLS-1192-1224 CLS-1192-2430 CLS-1192-2436
30622-738CS 185.13 CAD
30622-738 30622-740 30622-742
Autoclave Bags, Chemglass
Bags Sterilization Pouches
These durable 0.002'' thick transparent polypropylene autoclave bags provides maximum protection and performance for autoclavability.
  • These bags can hold 65, 200 and 240 standard size petri dishes
  • Steam autoclavable at 135 °C
It is used to sterlize labware such as Petri dishes, membrane filters, culture plates, pipettes, and more.