Rtx®-1 GC Columns, Restek
Supplier: Restek
10160 10183 10185 10192 10151 10153 10154 10156 10157 10158 10167 10170 10172 10173 10176 10178 10179 10179-126 10184 10188 10189 10193 10150 10155 10166 10169 10181 10182 10194 10253-124 12654-125 10152 10175 10177 10180 10190 10159 10187 10198 10154-125 10182-126 10180-600 10155-126 10167-126 10170-126 10180-125 10254-125 10157-128 10179-129 10185-129 10177-600 10178-600 10179-600 10173-129
RK10160EA 2675.24 CAD
RK10160 RK10183 RK10185 RK10192 RK10151 RK10153 RK10154 RK10156 RK10157 RK10158 RK10167 RK10170 RK10172 RK10173 RK10176 RK10178 RK10179 RK10179-126 RK10184 RK10188 RK10189 RK10191 RK10193 RK10150 RK10155 RK10166 RK10169 RK10181 RK10182 RK10194 RK10253-124 RK12654-125 RK10152 RK10175 RK10177 RK10180 RK10190 RK10159 RK10187 RK10198 10058-166 10058-168 10058-170 10058-172 10058-174 10058-176 10058-178 10058-180 10058-182 10058-184 10058-186 10058-190 10058-192 10058-194 10058-196 10058-198 10803-456 10849-158 10849-172 10849-182 10849-220 10849-296 10849-336 10849-566 10849-580 10849-718 10849-854 10849-872 10850-280 10850-282 10850-284 10850-290 10850-320 10850-376 10850-612 10850-614 10850-616 10850-728 10850-732 10850-742 10850-744 10850-766 10850-768 10850-770 10850-772 10850-776 10850-780 10850-834 10850-838 10852-078 10852-080 10852-096 75850-022 75850-878
Rtx®-1 GC Columns, Restek
Chromatography Columns
These general-purpose columns are designed for solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semi-volatiles, pesticides, and oxygenates.
  • Low bleed levels
  • Longer column lifetime
  • Short conditioning times
Columns exhibit long lifetimes and very low bleed at high operating temperatures. A proprietary synthesis process eliminates residual catalysts that could cause degradation and increase bleed. Phase composition: crossbonded 100% polydimethylpolysiloxane. Equivalent to USP G1, G2, and G38 phases.