WHEATON® Screw Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216376 W216377 W216378 W219365 W216375 W219366
10068-202CS 1749.46 CAD
10068-202 10068-206 10068-210 10070-712 10070-714 10070-716
WHEATON® Screw Thread Vials, DWK Life Sciences
Vials
The tubular design provides excellent clarity and dimensional consistency from vial to vial.
  • Ideal for lyophilization
  • Alternative to serum bottles and vials
  • Heavy walled glass provides impact resistance
  • Screw thread design eliminates need for crimping tools
These screw thread vials fits with I-Loc™ closure or screw cap with thin flange stopper.