ZymoPURE II™ Plasmid Kits, Zymo Research
Supplier: Zymo Research
D4200 D4201 D4202 D4203 D4204
77001-396EA 662.26 CAD
77001-396 77001-466 77001-468 77001-470 77001-472
ZymoPURE II™ Plasmid Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Vaccine/transfection grade plasmid DNA in ≤20 minutes.
  • Fastest: Simple 20 minute Midi/Maxi preps (50 min Giga)
  • Highest Yield: 6× more plasmid
  • Ultra-Pure: EndoZero™, vaccine grade and transfection ready