Assay Plates, 96-Well, Thomson Instrument
Supplier: THOMSON INSTRUMENT CO
931137 931512B 931130 931133 9356045
76808-480CS 458.91 CAD
76808-480 76808-492 76808-476 76808-478 76808-488
Assay Plates, 96-Well, Thomson Instrument
Microplates Deep Well Microplates
These 96-well assay plates are used for most common range of working volumes.
  • Made of LCMS tested PP
For long term storage use cap mat over a heat seal, but researchers have to be careful when defrosting. It is best to remove the capmat and replace with heat seal to avoid condensation raining into wells while thawing.