Peptide Synthesis Vessels, Solid Phase, Chemglass
Supplier: Chemglass
CG-1860-05 CG-1860-01 CG-1860-02 CG-1860-03 CG-1860-04 CG-1860-13 CG-1860-10 CG-1860-14 CG-1860-11 CG-1860-15 CG-1860-06 CG-1860-12
80071-356EA 711.67 CAD
80071-356 80071-348 80071-350 80071-352 80071-354 10170-980 10170-984 10170-988 10170-994 10170-998 10171-000 10171-004
Peptide Synthesis Vessels, Solid Phase, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Solid phase peptide synthesis vessel with a medium or coarse porosity fritted glass resin support.
Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Ordering information: Vessels are available in sizes other than those listed.