Genomic DNA Clean & Concentrator™ Kits, Zymo Research

Supplier: Zymo Research

D4010 D4011 D4064 D4065 D4066 D4067
77001-148EA 184.42 CAD
77001-148 77001-150 77001-340 77001-342 77001-344 77001-346
Genomic DNA Clean & Concentrator™ Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Quick (5 minute) clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.


  • Quick (5 minute) protocol
  • Unique spin column for low volume (≥ 10 µl) elution of ultra-pure, high-yield DNA
  • Eluted DNA is ideal for PCR, endonuclease digestion, sequencing, and more


The Genomic DNA Clean & Concentrator™ (DCC™) is designed for the quick (5 minute) recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga) DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations. Simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin™ Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, and more.

Order Now

SPECIFICATIONS

Description Genomic DNA Clean & Concentrator™ ZR-96 Genomic DNA Clean & Concentrator™
Environmentally Preferable
Application
Target molecule DNA DNA
Refine target molecule Amplified/modified DNA Amplified/modified DNA
Sample type PCR and other enzymatic reactions PCR and other enzymatic reactions
Method / Format Column (silica-based) Multiwell plate
Prep size Mini Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR