Potassium carbonate 15% (w/v)

Supplier: Ricca Chemical

Synonyms: Potash

R5883150-500A
RCR5883150500AEA 346.13 CAD
RCR5883150500A
Potassium carbonate 15% (w/v)
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: ∼100 °C
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://ca.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg

Order Now

Specification Test Results

Appearance Colorless Liquid
Physical State Liquid
pH 12
Initial Boiling Point /Range Approximately 100 °C
Relative Density 1.13
Solubility Miscible
Acute Toxicity LD50, Oral, Rat: 1870 mg/kg (Potassium Carbonate), details of toxic effects not reported other than lethal dose value

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR