2D Labtainer™ Bio Process Containers, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
SS00158-I SS00159-I SS00160-I SS00161-I SS00162-I SS00163-I SS00164-I
89259-736CS 5310.71 CAD
89259-736 89259-738 89259-740 89259-742 89259-744 89259-746 89259-748
2D Labtainer™ Bio Process Containers, Advanced Scientifics
Bags Media Bags
2D Labtainer™ bioProcess container (BPC) 2D system is composed of ASI™ 26/77 polyethylene film, a single-web, multi-layer 12.5 ml film.
  • Constructed in an ISO 7 clean room under cGMP conditions
  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
Applications:
Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
Preparing and storing buffer, culture media, and process liquids
Collecting chromatography feeds and fractions
Harvesting, storing, and transporting bulk drug product and drug precursors