EMS Holey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HC300-CU HC200-CU HC300-NI HC200-AU-25 HC300-AU-25 HC200-CU-100 HC300-CU-100 HC200-CU-150 HC200-NI-100 HC400-CU-150 HC200-AU-UL HC200-CU-UL HC200-NI-UL HC300-AU-UL HC300-CU-UL HC300-NI-UL HC400-AU-UL HC400-CU-UL
100501-076BX 457.11 CAD
100501-076 100501-072 100501-078 102101-350 102101-352 103305-566 103305-568 103305-572 103305-574 103305-580 76439-744 76439-752 76439-760 76439-768 76439-770 76439-778 76439-786 76439-794 76439-802 76439-810
EMS Holey Carbon Film, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of carbon with round holes is laid on the shiny side of the grid.
  • Coated grids are optically checked followed by batch testing in the EM
Available on copper, nickel, and gold 200, 300, and 400 mesh grids.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.