Screw Thread Culture Tubes, Kimble Chase
Supplier: DWK Life Sciences
90H13100R 90K16125RW 90P20125R 90K16125R
89236-730CS 628.36 CAD
89236-730 89236-724 89236-726 89236-728
Screw Thread Culture Tubes, Kimble Chase
Tubes Test Tubes
These borosilicate culture tubes are suitable for tissue culture work and general bacteriological use.
  • Round bottom and supplied without caps
  • No marking spot
Modular trays are shrink-wrapped in plastic film.