SP Wilmad-LabGlass 90° Bend Adapters with Stopcock, SP Industries
Supplier: SP Industries
LG-1710-102 ML-210-700
14208-004EA 235.85 CAD
14208-004 14210-400 14210-424 14210-526 14210-532 14210-600 14210-632 14210-834
SP Wilmad-LabGlass 90° Bend Adapters with Stopcock, SP Industries
Adapters Adapters, Joints with Tube Connectors
Made of borosilicate glass, these adapters have a 90° bend to 5/16" O.D. hose connection on top and inner Standard Taper joint on bottom.
Equipped with a stopcock, they are used to control gas/liquid addition in reaction systems.
Ordering information: ML-210 and LG-1710 have glass plugs, ML-211T and LG-1710T have PTFE plugs.