Fusidic acid 97.5-101.0% (by anhydrous basis), powder

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoic acid, Fusidine

26504-1G 26504-5G
CH26504-1GEA 55.06 CAD
CH26504-1G CH26504-5G
Fusidic acid 97.5-101.0% (by anhydrous basis), powder
Fusidic acid
Formula: C₃₁H₄₈O₆
MW: 516.72 g/mol
MDL Number: MFCD00865135
CAS Number: 6990-06-3

Order Now

Specification Test Results

Appearance White or off-white crystalline powder.
Solubility Practically insoluble in water, freely soluble in alcohol
Water Content (Karl Fisher) ≤2%
Infrared Spectrum Conforms to structure
Residual Solvent methanol ≤0.3%
Residual Solvent butyl acetate ≤ 0.1%
Residual Solvent butanol ≤ 0.05%
Total Impurities (related substances) ≤ 2%
Sulfated Ash ≤ 2%
Assay (anhydrous basis) 97.5 - 101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR