Staticmaster® EZ Mounts, NRD
Supplier: NRD Staticmaster
BF3 BF4
10183-838EA 105.45 CAD
10183-838 10183-840
Staticmaster® EZ Mounts, NRD
Ionizers
An indispensable accessory for mounting a 2U500 or 1U400 Ionizing Cartridge in the balance chamber.
  • Simple to use
  • Makes installation of ionizing cartridges fast and easy
Simply remove the protective layer of the double-sided tape and affix the E-Z mount to the back wall of the chamber. Once the mount is attached, installing ionizing cartridges is fast and easy.