TC100 Immersion Coolers, HUBER
Supplier: Huber
3005.0131.00 3005.0132.00
10221-070EA 31840.84 CAD
10221-070 10221-072 10221-074 10221-076
TC100 Immersion Coolers, HUBER
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Huber Immersion Coolers are easy to use and ideally suited to quick liquid cooling.
  • Universal solution for many cooling tasks
  • Rapid cooling of fluids
  • Ideal for counter-cooling or as a replacement of dry ice
  • LED temperature display
  • Easy operation with only three keys
The TC range of Immersion Coolers are a flexible solution for a variety of cooling tasks
The models with the type affix "E“ have an LED display with entry of nominal values and regulate with a precision of ±0.5 K. A spiral-shaped or flexible immersion cooling probe made out ofMore Product Information