Coenzyme A sodium salt hydrate ≥98%

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: CoA Na2, COA NA2 Hydrate

01534-100MG 01534-250MG 01534-25MG
CH01534-100MGEA 829.5 CAD
CH01534-100MG CH01534-250MG CH01534-25MG
Coenzyme A sodium salt hydrate ≥98%
Coenzyme A sodium salt hydrate

Enzymatically assayed using phosphotransacetylase. Approx. 95% CoA-SH

Formula: C₂₁H₃₆N₇O₁₆P₃S·xNa⁺·yH₂O
MW: 767.53 g/mol (free acid, anhy)
MDL Number: MFCD06795857
CAS Number: 55672-92-9

Order Now

Specification Test Results

Purity Limit ≥ 98%
Purity Method HPLC, UV Method
Molecular Weight 807.52
Appearance Amorphous light creamy powder
Storage Temperature Store at ≤ −4 °C
Solubility Dissolves in cold water
Odor Typical thiol-group odor
Assay ≤ 90% (Phosphotransacetylase)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR