VWR® Bead Sterilizers
Supplier: VWR International
75999-324 75999-328 75999-332
75999-324EA 997.95 CAD
75999-324 75999-328 75999-332
VWR® Bead Sterilizers
Sterilizers Glass Bead Sterilizers
Research sterilizer for needles, forceps, tweezers, and other laboratory tools.
  • Digital control of temperature up to 300 °C eliminates the need for flame sterilization
The VWR® Micro Sterilizer is a fast and efficient instrument for sterilizing small research tools including forceps, needles, and tweezers. With a powerful heating element that can quickly reach temperatures as high as 300 °C, this bead sterilizer can reduce the presence of bacteria, spores, and micro-organisms. The unit's chamber temperature is viewed on a large LED panel and heat can be controlled using the control knob. Also available as an "XL" model for sterilizing larger/taller tools.
Certifications: CE marked.
Accessories information: Extra glass beads are available separately.