Glycerine ≥99.5%, AnalaR NORMAPUR® ACS, redistilled, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Propane-1,2,3-triolGlycerol1,2,3-Propanetriol

24388.320
BDH24388.320CS 1340.58 CAD
BDH24388.320
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS, redistilled, VWR Chemicals BDH®
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://ca.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Assay Min. 99.5 %
Acrolein and glucose Passes test
Appearance of solution (10 % V/V; water) Passes test
Substances darkened by sulphuric acid Passes test
Acidity Max. 0.0004 meq/g
Alkalinity Max. 0.0003 meq/g
Colouration Max. 10 APHA
Density (20/4) 1.257 - 1.265
Acetaldehyde Max. 10 ppm
Esters of fatty acids (as C15H26O6) Max. 0.05 %
Glyceraldehyde Max. 30 ppm
Heavy metals (as Pb) Max. 1 ppm
Ignition residue (SO4) Max. 50 ppm
Ketones (as CH3COCH3) Max. 50 ppm
Water Max. 0.5 %
Cl (Chloride) Max. 1 ppm
NH4 (Ammonium) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
As (Arsenic) Max. 1 ppm
Ba (Barium) Max. 5 ppm
Fe (Iron) Max. 1 ppm
Conforms to ACS Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR