illustra NAP Columns, Cytiva
Supplier: Cytiva
17-0852-02 17-0852-01 17-0853-01 17-0853-02 17-0854-01 17-0854-02
CA95017-005LPK 1547.38 CAD
CA95017-005L CA95017-003L CA95017-007L CA95017-009L CA95017-011L CA95017-013L
illustra NAP Columns, Cytiva
Chromatography Columns
NAP Columns are disposable columns, prepacked with Sephadex G-25 DNA Grade, that require only gravity to run.
  • For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flow
  • Prepacked with Sephadex G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide
  • Available in three sizes depending on sample volume: 0.5 ml (NAP-5), 1 ml (NAP-10) or 2.5 ml (NAP-25)
  • Useful for small-scale purification, desalting and buffer exchange
Delivery information: Each package includes a complete step-by-stepMore Product Information