PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
1304004001-5F 1304005005-5F 1304006002-5F 1301005001-10F 1301005001-5F 1301007002-5F 1301010003-5F 1301015008-5F 1301020004-5F 1302005001-5F 1302007002-5F 1302010003-5F 1302015008-5F 1302020004-5F 1302030005-5F 1302040000-5F 1303062000-5F 1303080000-5F 1304002000-5F 1304003006-5F 1301007002-10F 1301010003-10F 1301015008-10F 1301020004-10F 1302005001-10F 1302007002-10F 1302010003-10F 1302015008-10F 1302020004-10F 1302030005-10F 1302040000-10F 1303062000-10F 1303080000-10F 1304002000-10F 1304003006-10F 1304004001-10F 1304005005-10F 1304006002-10F 1301005001-50F 1301007002-50F 1301010003-50F 1301015008-50F 1301020004-50F 1302005001-50F 1302007002-50F 1302010003-50F 1302015008-50F 1302020004-50F 1302030005-50F 1302040000-50F 1303062000-50F 1303080000-50F 1304002000-50F 1304003006-50F 1304004001-50F 1304005005-50F 1304006002-50F 1301005001-100F 1301007002-100F 1301010003-100F 1301015008-100F 1301020004-100F 1302005001-100F 1302007002-100F 1302010003-100F 1302015008-100F 1302020004-100F 1302030005-100F 1302040000-100F 1303062000-100F 1303080000-100F 1304002000-100F 1304003006-100F 1304004001-100F 1304005005-100F 1304006002-100F 1302013004-5F 1302030001-5F 1302013004-10F 1302030001-10F 1302013004-50F 1302030001-50F 1302013004-100F 1302030001-100F 1302004006-5F 1302004006-10F 1302004006-50F 1302004006-100F
76151-844EA 84.47 CAD
76151-844 76151-846 76151-848 76151-860 76151-862 76151-864 76151-866 76151-868 76151-870 76151-872 76151-874 76151-876 76151-878 76151-880 76151-882 76151-884 76151-886 76151-888 76151-890 76151-892 76151-894 76151-896 76151-898 76151-900 76151-902 76151-904 76151-906 76151-908 76151-910 76151-912 76151-914 76151-916 76151-918 76151-920 76151-922 76151-924 76151-926 76151-928 76151-930 76151-932 76151-934 76151-936 76151-938 76151-940 76151-942 76151-944 76151-946 76151-948 76151-950 76151-952 76151-954 76151-956 76151-958 76151-960 76151-962 76151-964 76151-966 76151-968 76151-970 76151-972 76151-974 76151-976 76151-978 76151-980 76151-982 76151-984 76151-986 76151-988 76151-990 76151-992 76151-994 76151-996 76151-998 76152-000 76152-002 76152-004 76152-006 76152-008 76152-010 76152-012 76152-014 76152-016 76152-018 76152-020 76152-022 76152-024 76152-026 76152-028
PEEK Tubing, Trajan Scientific and Medical
Tubing Chromatography Tubing
Trajan precision bore PEEK tubing is designed specifically for modern LC, LCMS, and automation usage.
  • Excellent chemical resistance and inert to most commonly used solvents
  • Bio-inert and bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with polymer and stainless steel fittings
  • Can be formed to specific customer needs
Trajan precision bore PEEK tubing ensures reproducibility in chromatographic flow rates and results through smooth internalMore Product Information