PTFE-Lined Closed Caps, Chemglass
Supplier: Chemglass
CG-895-02 CG-895-03 CG-895-04 CG-895-07
10170-362PK 126.97 CAD
10170-362 10170-780 10170-782 10170-784
PTFE-Lined Closed Caps, Chemglass
Closures Bottle Closures
Caps features foam backed PTFE that resists attack from virtually all chemicals at room temperature, while providing a leak-tight seal.
  • Temperature tolerant
  • Not autoclavable