pH Indicator Papers, Duotest, Machery Nagel
Supplier: Macherey-Nagel
90303 90304 90305 90301
89049-908EA 51.87 CAD
89049-908 89049-914 89049-916 89049-912
pH Indicator Papers, Duotest, Machery Nagel
Test Papers pH Indicator Papers and Test Strips
Duotest indicator papers combine two different indicator zones on a single strip
The zones are separated by a hydrophobic barrier, which effectively prevents mixing of the reaction colors and increases the mechanical stability.